Assay Details
Target Gene Details
Entrez Gene ID: | 51338 |
Gene Name: | membrane spanning 4-domains A4A |
Gene Aliases: |
4SPAN1, CD20-L1, CD20L1, HDCME31P, MS4A4, MS4A7 |
Location: |
Chr.11:60280541-60308972 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A4A | NM_001243266.1 | NP_001230195.1 | ||
| NM_024021.3 | NP_076926.2 | |||
| NM_148975.2 | NP_683876.1 | |||
| XM_017017909.1 | 1 | 108 | XP_016873398.1 | |
| AB013102.1 | BAB18738.1 | |||
| AB022821.1 | BAB61018.1 | |||
| AF068288.1 | AAF65507.1 | |||
| AF237912.1 | AAK37594.1 | |||
| AK075081.1 | BAC11389.1 | |||
| AK292089.1 | ||||
| AK314033.1 | ||||
| BC020648.1 | AAH20648.1 | |||
| BG538851.1 | ||||
| CB046393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832174 | Chr.11:60141794 - 60306435 on Build GRCh38 | Gain |
|
| nsv1053558 | Chr.11:60211987 - 60293736 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map