Assay Details
Target Gene Details
Entrez Gene ID: | 2620 |
Gene Name: | growth arrest specific 2 |
Gene Aliases: |
- |
Location: |
Chr.11:22626113-22885930 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS2 | NM_001143830.1 | NP_001137302.1 | ||
| NM_177553.2 | NP_808221.1 | |||
| XM_011519971.2 | XP_011518273.1 | |||
| XM_011519972.2 | XP_011518274.1 | |||
| XM_011519975.1 | XP_011518277.1 | |||
| XM_011519978.2 | XP_011518280.1 | |||
| XM_017017528.1 | XP_016873017.1 | |||
| XM_017017529.1 | XP_016873018.1 | |||
| XM_017017530.1 | XP_016873019.1 | |||
| XM_017017531.1 | XP_016873020.1 | |||
| XM_017017532.1 | XP_016873021.1 | |||
| AK313734.1 | ||||
| BC013326.1 | AAH13326.1 | |||
| BC040470.1 | AAH40470.1 | |||
| DA345287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046773 | Chr.11:22597901 - 23427777 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75718428] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map