Assay Details
Target Gene Details
Entrez Gene ID: | 4041 |
Gene Name: | LDL receptor related protein 5 |
Gene Aliases: |
BMND1, EVR1, EVR4, HBM, LR3, LRP-5, LRP7, OPPG, OPS, OPTA1, VBCH2 |
Location: |
Chr.11:68298866-68449275 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP5 | NM_001291902.1 | NP_001278831.1 | ||
| NM_002335.3 | NP_002326.2 | |||
| XM_005273994.2 | XP_005274051.1 | |||
| XM_011545029.1 | XP_011543331.1 | |||
| XM_011545030.1 | XP_011543332.1 | |||
| XM_011545031.1 | XP_011543333.1 | |||
| AB017498.1 | BAA33051.1 | |||
| AF064548.1 | AAC36467.1 | |||
| AF077820.1 | AAC72791.1 | |||
| AF088027.1 | ||||
| BC150595.1 | ||||
| DN999254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825963 | Chr.11:67996205 - 68429586 on Build GRCh38 | Gain |
|
| esv2656496 | Chr.11:68301829 - 68466947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map