Assay Details
Target Gene Details
Entrez Gene ID: | 7054 |
Gene Name: | tyrosine hydroxylase |
Gene Aliases: |
DYT14, DYT5b, TYH |
Location: |
Chr.11:2163929-2174081 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TH | XM_011520335.2 | 1 | 349 | XP_011518637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553070 | Chr.11:2154812 - 2177188 on Build GRCh38 | Loss |
|
| nsv469927 | Chr.11:2165105 - 2201683 on Build GRCh38 | Loss |
|
| nsv553068 | Chr.11:2096173 - 2174751 on Build GRCh38 | Loss |
|
| nsv951284 | Chr.11:2083671 - 2218670 on Build GRCh38 | Deletion |
|
| nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73403756] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map