Assay Details
Target Gene Details
Entrez Gene ID: | 84953 |
Gene Name: | MICAL C-terminal like |
Gene Aliases: |
Ebitein1 |
Location: |
Chr.11:12286900-12359144 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICALCL | NM_032867.2 | NP_116256.2 | ||
| AK027872.1 | BAB55422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625390 | Chr.11:12320996 - 12405629 on Build GRCh38 | Gain |
|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| dgv1059n100 | Chr.11:12197656 - 12685051 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map