Assay Details
Target Gene Details
Entrez Gene ID: | 2348 |
Gene Name: | folate receptor 1 |
Gene Aliases: |
FBP, FOLR |
Location: |
Chr.11:72189558-72196323 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOLR1 | NM_000802.3 | 2 | 70 | NP_000793.1 |
| NM_016724.2 | 3 | 321 | NP_057936.1 | |
| NM_016725.2 | 2 | 255 | NP_057937.1 | |
| NM_016729.2 | 1 | 90 | NP_057941.1 | |
| AK223527.1 | 2 | 109 | BAD97247.1 | |
| BC002947.2 | 2 | 202 | AAH02947.1 | |
| BT007158.1 | 1 | 45 | AAP35822.1 | |
| CR542019.1 | 1 | 45 | CAG46816.1 | |
| J05013.1 | 2 | 256 | AAA35823.1 | |
| KU257686.1 | ||||
| M25317.1 | AAA74896.1 | |||
| M28099.1 | 3 | 141 | AAA35822.1 | |
| M35069.1 | AAA35824.1 | |||
| U78793.1 | 1 | 90 | AAB39751.1 | |
| U78794.1 | 1 | 71 | AAB39752.1 | |
| X62753.1 | 2 | 475 | CAA44610.1 |
More Information
Set Membership: |
|

Genomic Map