Assay Details
Target Gene Details
Entrez Gene ID: | 159963 |
Gene Name: | solute carrier family 5 member 12 |
Gene Aliases: |
SMCT2 |
Location: |
Chr.11:26667019-26723356 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A12 | NM_178498.3 | 15 | 4710 | NP_848593.2 |
| XM_006718155.3 | 15 | 4613 | XP_006718218.1 | |
| XM_011519920.2 | 16 | 4190 | XP_011518222.1 | |
| XM_017017244.1 | 16 | 4636 | XP_016872733.1 | |
| AL049270.1 | 1 | 705 |
More Information
Set Membership: |
|

Genomic Map