Assay Details
Target Gene Details
Entrez Gene ID: | 399959 |
Gene Name: | mir-100-let-7a-2 cluster host gene |
Gene Aliases: |
AGD1, linc-NeD125, lncRNA-N2 |
Location: |
Chr.11:122089103-122203062 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR100HG | NR_024430.2 | 4 | 651 | |
| NR_137175.1 | 2 | 922 | ||
| NR_137176.1 | 2 | 633 | ||
| NR_137177.1 | 2 | 612 | ||
| NR_137178.1 | ||||
| NR_137179.1 | 5 | 605 | ||
| NR_137180.1 | 5 | 663 | ||
| NR_137181.1 | 3 | 548 | ||
| NR_137182.1 | 4 | 630 | ||
| NR_137183.1 | 3 | 539 | ||
| NR_137184.1 | 2 | 457 | ||
| NR_137185.1 | 3 | 518 | ||
| NR_137186.1 | 3 | 427 | ||
| NR_137187.1 | 3 | 406 | ||
| NR_137188.1 | 3 | 496 | ||
| NR_137189.1 | 2 | 943 | ||
| NR_137190.1 | 3 | 976 | ||
| NR_137191.1 | 3 | 955 | ||
| NR_137192.1 | 4 | 815 | ||
| NR_137193.1 | 5 | 642 | ||
| NR_137194.1 | 3 | 517 | ||
| NR_137195.1 | 2 | 685 | ||
| NR_137196.1 | 2 | 706 | ||
| AB075498.1 | 2 | 1175 | ||
| AB485963.1 | 4 | 533 | ||
| AK091713.1 | 3 | 307 | ||
| AK123947.1 | 2 | 859 | ||
| AK311309.1 | 4 | 604 | ||
| AL832469.1 | 2 | 239 | ||
| BX538058.1 | 2 | 212 | ||
| BX647608.1 | 2 | 213 | ||
| DA027094.1 | 3 | 350 | ||
| DA153922.1 | ||||
| DA349788.1 | 3 | 358 | ||
| DA474693.1 | ||||
| DA532588.1 | 3 | 430 | ||
| DA597554.1 | 3 | 427 | ||
| DA649890.1 | 3 | 302 | ||
| DA650430.1 | ||||
| DA659486.1 | 2 | 367 | ||
| DA769636.1 | 2 | 472 | ||
| DA788103.1 | ||||
| DA861327.1 | ||||
| DB248200.1 | 3 | 325 |

Genomic Map