Assay Details
Target Gene Details
Entrez Gene ID: | 1075 |
Gene Name: | cathepsin C |
Gene Aliases: |
CPPI, DPP-I, DPP1, DPPI, HMS, JP, JPD, PALS, PDON1, PLS |
Location: |
Chr.11:88293592-88337787 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSC | NM_001814.5 | NP_001805.3 | ||
| AF234263.1 | AAL48191.1 | |||
| AF234264.1 | AAL48192.1 | |||
| AF254757.1 | AAL48195.1 | |||
| AF525032.1 | AAQ08887.1 | |||
| AF525033.1 | AAQ08888.1 | |||
| AK223038.1 | BAD96758.1 | |||
| AK292117.1 | ||||
| AK296190.1 | ||||
| BX380095.2 | ||||
| BX459226.2 | ||||
| X87212.1 | CAA60671.1 |
More Information
Set Membership: |
|

Genomic Map