Assay Details
Target Gene Details
Entrez Gene ID: | 143884 |
Gene Name: | CWF19-like 2, cell cycle control (S. pombe) |
Gene Aliases: |
- |
Location: |
Chr.11:107326345-107457846 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWF19L2 | NM_152434.2 | NP_689647.2 | ||
| XM_017017239.1 | XP_016872728.1 | |||
| AK056905.1 | BAB71307.1 | |||
| AK126881.1 | ||||
| AK293001.1 | ||||
| BC056241.1 | AAH56241.1 | |||
| BC110439.1 | ||||
| BC110440.1 | ||||
| BC110441.1 | ||||
| BC110442.1 | ||||
| BC118669.1 | ||||
| BX647832.1 | ||||
| BX648704.1 | ||||
| BX649022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627644 | Chr.11:107257370 - 107361314 on Build GRCh38 | Gain |
|
| nsv556281 | Chr.11:107327143 - 107370076 on Build GRCh38 | Loss |
|
| nsv556280 | Chr.11:107288014 - 107360879 on Build GRCh38 | Gain |
|
| nsv832261 | Chr.11:107275175 - 107450836 on Build GRCh38 | Loss |
|
| nsv475 | Chr.11:107341074 - 107384996 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map