Assay Details
Target Gene Details
Entrez Gene ID: | 8550 |
Gene Name: | mitogen-activated protein kinase-activated protein kinase 5 |
Gene Aliases: |
MAPKAP-K5, MK-5, MK5, PRAK |
Location: |
Chr.12:111842228-111902225 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAPK5 | NM_003668.3 | NP_003659.2 | ||
| NM_139078.2 | NP_620777.1 | |||
| XM_017020125.1 | XP_016875614.1 | |||
| XM_017020126.1 | XP_016875615.1 | |||
| XM_017020127.1 | XP_016875616.1 | |||
| XM_017020128.1 | XP_016875617.1 | |||
| XM_017020129.1 | XP_016875618.1 | |||
| XM_017020130.1 | XP_016875619.1 | |||
| XM_017020131.1 | XP_016875620.1 | |||
| XM_017020132.1 | XP_016875621.1 | |||
| XM_017020133.1 | XP_016875622.1 | |||
| XM_017020134.1 | XP_016875623.1 | |||
| XM_017020135.1 | XP_016875624.1 | |||
| XM_017020136.1 | XP_016875625.1 | |||
| XM_017020137.1 | XP_016875626.1 | |||
| XM_017020139.1 | XP_016875628.1 | |||
| AK122767.1 | ||||
| AW780199.1 | ||||
| BC000833.1 | AAH00833.1 | |||
| BC041049.1 | AAH41049.1 | |||
| BC047284.1 | AAH47284.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760710 | Chr.12:111735404 - 111876910 on Build GRCh38 | Gain |
|
| dgv514e212 | Chr.12:111735392 - 111884067 on Build GRCh38 | Gain |
|
| dgv304e214 | Chr.12:111742954 - 111877939 on Build GRCh38 | Gain |
|
| nsv470317 | Chr.12:111761276 - 111868361 on Build GRCh38 | Gain |
|
| esv3630770 | Chr.12:111861669 - 111872939 on Build GRCh38 | Gain |
|
| dgv1556n100 | Chr.12:111732560 - 111876898 on Build GRCh38 | Gain |
|
| dgv2874n54 | Chr.12:111743274 - 111884680 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map