Assay Details
Target Gene Details
Entrez Gene ID: | 440145 |
Gene Name: | mitotic spindle organizing protein 1 |
Gene Aliases: |
C13orf37, MOZART1 |
Location: |
Chr.13:72708357-72727800 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MZT1 | NM_001071775.2 | NP_001065243.1 | ||
| AK025522.1 | ||||
| BC125183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3632699 | Chr.13:72725451 - 72726164 on Build GRCh38 | Loss |
|
| esv2667348 | Chr.13:72725459 - 72726167 on Build GRCh38 | Deletion |
|
| esv2747634 | Chr.13:72725468 - 72726339 on Build GRCh38 | Deletion |
|
| nsv826707 | Chr.13:72725428 - 72726244 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map