Assay Details
Target Gene Details
Entrez Gene ID: | 2308 |
Gene Name: | forkhead box O1 |
Gene Aliases: |
FKH1, FKHR, FOXO1A |
Location: |
Chr.13:40555664-40666597 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXO1 | NM_002015.3 | NP_002006.2 | ||
| XM_011535008.2 | XP_011533310.1 | |||
| XM_011535010.2 | XP_011533312.1 | |||
| AF032885.1 | AAC39591.1 | |||
| AK310511.1 | ||||
| BC021981.2 | AAH21981.1 | |||
| BC070065.1 | ||||
| BT007455.1 | AAP36123.1 | |||
| BX648278.1 | ||||
| U02310.1 | AAA03629.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525547 | Chr.13:40550515 - 40563667 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map