Assay Details
Target Gene Details
Entrez Gene ID: | 8803 |
Gene Name: | succinate-CoA ligase ADP-forming beta subunit |
Gene Aliases: |
A-BETA, MTDPS5, SCS-betaA |
Location: |
Chr.13:47942656-48037972 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUCLA2 | NM_003850.2 | NP_003841.1 | ||
| XM_017020806.1 | XP_016876295.1 | |||
| XM_017020807.1 | XP_016876296.1 | |||
| XM_017020809.1 | XP_016876298.1 | |||
| AB035863.1 | BAA92873.1 | |||
| AF058953.1 | AAC64396.1 | |||
| AF087890.1 | AAP97189.1 | |||
| AK001458.1 | BAA91703.1 | |||
| AK001847.1 | BAA91939.1 | |||
| AK093506.1 | ||||
| AK225452.1 | ||||
| AK299437.1 | ||||
| AK302311.1 | ||||
| AK315513.1 | ||||
| AK316041.1 | ||||
| AK316271.1 | ||||
| AL080096.2 | CAB45708.2 | |||
| BC027587.2 | AAH27587.1 | |||
| BX640602.1 | CAE45708.1 | |||
| KU178442.1 | ||||
| KU178443.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76263305] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map