Assay Details
Target Gene Details
Entrez Gene ID: | 100874222 |
Gene Name: | RNF219 antisense RNA 1 |
Gene Aliases: |
POU4F1-AS1 |
Location: |
Chr.13:78054855-78617325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF219-AS1 | NR_047001.1 | |||
| AK095779.1 | ||||
| BX647243.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73233136] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map