Assay Details
Target Gene Details
Entrez Gene ID: | 5095 |
Gene Name: | propionyl-CoA carboxylase alpha subunit |
Gene Aliases: |
- |
Location: |
Chr.13:100089015-100530437 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832698 | Chr.13:100068689 - 100238635 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map