Assay Details
Target Gene Details
Entrez Gene ID: | 64062 |
Gene Name: | RNA binding motif protein 26 |
Gene Aliases: |
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17 |
Location: |
Chr.13:79311827-79406265 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM26 | XM_005266491.4 | 24 | 7302 | XP_005266548.1 |
| XM_005266497.1 | 22 | 6067 | XP_005266554.1 | |
| XM_006719857.1 | 22 | 6064 | XP_006719920.1 | |
| XM_011535180.2 | 24 | 7299 | XP_011533482.1 | |
| XM_011535183.2 | 24 | 7293 | XP_011533485.1 | |
| XM_011535185.2 | 24 | 7272 | XP_011533487.1 | |
| XM_011535187.2 | 24 | 7263 | XP_011533489.1 | |
| XM_011535188.2 | 24 | 7264 | XP_011533490.1 | |
| XM_011535191.2 | 23 | 7221 | XP_011533493.1 | |
| XM_011535192.2 | 23 | 7192 | XP_011533494.1 | |
| XM_011535193.2 | 23 | 7176 | XP_011533495.1 | |
| XM_017020688.1 | 24 | 7299 | XP_016876177.1 | |
| XM_017020690.1 | 24 | 7294 | XP_016876179.1 | |
| XM_017020691.1 | 24 | 7285 | XP_016876180.1 | |
| XM_017020692.1 | 24 | 7255 | XP_016876181.1 | |
| XM_017020693.1 | 23 | 7230 | XP_016876182.1 | |
| XM_017020694.1 | 23 | 7215 | XP_016876183.1 | |
| XM_017020697.1 | 22 | 6056 | XP_016876186.1 | |
| XM_017020699.1 | 23 | 7207 | XP_016876188.1 | |
| XM_017020701.1 | 24 | 7360 | XP_016876190.1 | |
| XM_017020702.1 | 22 | 6125 | XP_016876191.1 | |
| XM_017020703.1 | 24 | 7330 | XP_016876192.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3632816 | Chr.13:79199236 - 79375478 on Build GRCh38 | Gain |
|
| esv3632814 | Chr.13:79199135 - 79520919 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map