Assay Details
Target Gene Details
Entrez Gene ID: | 55757 |
Gene Name: | UDP-glucose glycoprotein glucosyltransferase 2 |
Gene Aliases: |
HUGT2, UGCGL2, UGT2 |
Location: |
Chr.13:95801580-96053482 on Build GRCh38 |
Assay Gene Location: | Within Intron 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGGT2 | NM_020121.3 | NP_064506.3 | ||
| XM_011521094.1 | XP_011519396.1 | |||
| XM_011521095.1 | XP_011519397.1 | |||
| XM_011521096.1 | XP_011519398.1 | |||
| XM_011521097.1 | XP_011519399.1 | |||
| XM_011521098.1 | XP_011519400.1 | |||
| XM_011521099.2 | XP_011519401.1 | |||
| XM_011521102.2 | XP_011519404.1 | |||
| XM_011521103.2 | XP_011519405.1 | |||
| XM_017020662.1 | XP_016876151.1 | |||
| XM_017020663.1 | XP_016876152.1 | |||
| AF227906.2 | AAF66233.2 | |||
| AL133051.1 | CAB61378.1 | |||
| BC125233.1 | ||||
| BC125234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832691 | Chr.13:95825844 - 96007569 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114507333] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map