Assay Details
Target Gene Details
Entrez Gene ID: | 171425 |
Gene Name: | citrate lyase beta like |
Gene Aliases: |
CLB |
Location: |
Chr.13:99606664-99909459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLYBL | NM_206808.3 | NP_996531.1 | ||
| NR_104592.1 | ||||
| XM_005254030.2 | XP_005254087.1 | |||
| XM_005254031.2 | XP_005254088.1 | |||
| XM_006719915.3 | XP_006719978.1 | |||
| XM_011521051.2 | XP_011519353.1 | |||
| XM_011521052.2 | XP_011519354.1 | |||
| XM_017020402.1 | XP_016875891.1 | |||
| XM_017020403.1 | XP_016875892.1 | |||
| AF428253.1 | AAL84703.1 | |||
| AK095506.1 | BAC04561.1 | |||
| AK300510.1 | ||||
| BC034360.1 | AAH34360.1 | |||
| DA621336.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34003 | Chr.13:99451122 - 99636703 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74112517] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map