Assay Details
Target Gene Details
Entrez Gene ID: | 7223 |
Gene Name: | transient receptor potential cation channel subfamily C member 4 |
Gene Aliases: |
HTRP-4, HTRP4, TRP4 |
Location: |
Chr.13:37633872-37870374 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34109 | Chr.13:37269682 - 37702254 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map