Assay Details
Target Gene Details
Entrez Gene ID: | 5911 |
Gene Name: | RAP2A, member of RAS oncogene family |
Gene Aliases: |
K-REV, KREV, RAP2, RbBP-30 |
Location: |
Chr.13:97434221-97467998 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2A | NM_021033.6 | NP_066361.1 | ||
| AF205602.1 | AAN71845.1 | |||
| AF493914.1 | AAM12628.1 | |||
| AK311202.1 | ||||
| AK315139.1 | ||||
| BC041333.2 | AAH41333.1 | |||
| BC070031.1 | AAH70031.1 | |||
| X12534.1 | CAA31052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832694 | Chr.13:97278307 - 97485628 on Build GRCh38 | Gain |
|
| dgv3332n54 | Chr.13:97446270 - 97456107 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map