Assay Details
Target Gene Details
Entrez Gene ID: | 407975 |
Gene Name: | miR-17-92a-1 cluster host gene |
Gene Aliases: |
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92 |
Location: |
Chr.13:91347820-91354575 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR17HG | NR_027349.1 | |||
| NR_027350.1 | 2 | 4418 | ||
| AB176707.1 | ||||
| AK024240.1 | 1 | 2141 | ||
| BC040320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832679 | Chr.13:91207213 - 91377966 on Build GRCh38 | Gain |
|

Genomic Map