Assay Details
Target Gene Details
Entrez Gene ID: | 23281 |
Gene Name: | microtubule associated tumor suppressor candidate 2 |
Gene Aliases: |
CAZIP, ICIS, KIAA0774, TIP150 |
Location: |
Chr.13:28820314-29505947 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTUS2 | XM_011535019.2 | XP_011533321.1 | ||
| XM_011535020.2 | XP_011533322.1 | |||
| XM_011535021.2 | XP_011533323.1 | |||
| XM_017020500.1 | XP_016875989.1 | |||
| XM_017020501.1 | XP_016875990.1 | |||
| XM_017020502.1 | XP_016875991.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73161836] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map