Assay Details
Target Gene Details
Entrez Gene ID: | 100288730 |
Gene Name: | PAN3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:28136843-28139174 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAN3-AS1 | NR_029383.1 | 2 | 1338 | |
| AK307165.1 | 2 | 1347 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34052 | Chr.13:28074532 - 28352440 on Build GRCh38 | Loss |
|

Genomic Map