Assay Details
Target Gene Details
Entrez Gene ID: | 407975 |
Gene Name: | miR-17-92a-1 cluster host gene |
Gene Aliases: |
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92 |
Location: |
Chr.13:91347820-91354575 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR17HG | NR_027349.1 | |||
| NR_027350.1 | ||||
| AB176707.1 | ||||
| BC040320.1 | ||||
| BC109081.2 | ||||
| BC109082.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3317n54 | Chr.13:91348005 - 91349218 on Build GRCh38 | Loss |
|
| esv28603 | Chr.13:91347381 - 91349183 on Build GRCh38 | Loss |
|
| dgv3313n54 | Chr.13:91339903 - 91349218 on Build GRCh38 | Loss |
|
| dgv3315n54 | Chr.13:91347886 - 91349096 on Build GRCh38 | Gain+Loss |
|
| nsv562697 | Chr.13:91347483 - 91348809 on Build GRCh38 | Loss |
|
| nsv832679 | Chr.13:91207213 - 91377966 on Build GRCh38 | Gain |
|

Genomic Map