Assay Details
Target Gene Details
Entrez Gene ID: | 3981 |
Gene Name: | DNA ligase 4 |
Gene Aliases: |
LIG4S |
Location: |
Chr.13:108207442-108218368 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIG4 | NM_001098268.1 | NP_001091738.1 | ||
| XM_005254058.3 | 1 | 1398 | XP_005254115.1 | |
| XM_006719951.3 | 1 | 1476 | XP_006720014.1 | |
| XM_006719952.1 | XP_006720015.1 | |||
| XM_011521091.2 | XP_011519393.1 | |||
| XM_017020563.1 | 1 | 1398 | XP_016876052.1 | |
| XM_017020565.1 | XP_016876054.1 | |||
| XM_017020566.1 | XP_016876055.1 | |||
| XM_017020568.1 | 1 | 1398 | XP_016876057.1 | |
| XM_017020569.1 | 2 | 1847 | XP_016876058.1 | |
| XM_017020570.1 | XP_016876059.1 | |||
| XM_017020571.1 | 2 | 1847 | XP_016876060.1 | |
| XM_017020572.1 | 1 | 1398 | XP_016876061.1 | |
| BX648833.1 |
More Information
Set Membership: |
|

Genomic Map