Assay Details
Target Gene Details
Entrez Gene ID: | 160851 |
Gene Name: | diacylglycerol kinase eta |
Gene Aliases: |
DGKeta |
Location: |
Chr.13:42040036-42256584 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKH | NM_001204504.2 | NP_001191433.1 | ||
| AK289546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832588 | Chr.13:41876984 - 42053116 on Build GRCh38 | Gain |
|
| nsv1016 | Chr.13:42041861 - 42072289 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map