Assay Details
Target Gene Details
Entrez Gene ID: | 55671 |
Gene Name: | protein phosphatase 4 regulatory subunit 3A |
Gene Aliases: |
FLFL1, KIAA2010, MSTP033, PP4R3, PP4R3A, SMEK1, smk-1, smk1 |
Location: |
Chr.14:91457481-91510554 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R3A | NM_001284280.1 | 15 | 4515 | NP_001271209.1 |
| NM_001284281.1 | 13 | 3837 | NP_001271210.1 | |
| AI608781.1 | 1 | 418 |
More Information
Set Membership: |
|

Genomic Map