Assay Details
Target Gene Details
Entrez Gene ID: | 161394 |
Gene Name: | sterile alpha motif domain containing 15 |
Gene Aliases: |
C14orf174, FAM15A |
Location: |
Chr.14:77377419-77391244 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD15 | NM_001010860.1 | NP_001010860.1 | ||
| AK292571.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77088756] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map