Assay Details
Target Gene Details
Entrez Gene ID: | 4625 |
Gene Name: | myosin, heavy chain 7, cardiac muscle, beta |
Gene Aliases: |
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM |
Location: |
Chr.14:23412738-23435686 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYH7 | NM_000257.3 | NP_000248.2 | ||
| XM_017021340.1 | XP_016876829.1 | |||
| AB209708.1 | BAD92945.1 | |||
| AK307590.1 | ||||
| AY518538.1 | AAT44534.1 | |||
| BC112171.1 | AAI12172.1 | |||
| BC112173.1 | AAI12174.1 | |||
| DQ248310.1 | ABB55265.1 | |||
| EF560725.1 | ||||
| EU747717.1 | ||||
| M58018.1 | AAA62830.1 | |||
| X51591.1 | CAA35940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584149 | Chr.14:23395407 - 23427942 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map