Assay Details
Target Gene Details
Entrez Gene ID: | 79178 |
Gene Name: | thiamine triphosphatase |
Gene Aliases: |
THTP, THTPASE |
Location: |
Chr.14:23511760-23559581 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THTPA | NR_046051.1 | |||
| NR_046052.1 | ||||
| AK090747.1 | ||||
| AK094211.1 | ||||
| BI488427.1 | ||||
| DA218228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564043 | Chr.14:23495437 - 23550918 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115840174] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map