Assay Details
Target Gene Details
Entrez Gene ID: | 122704 |
Gene Name: | mitochondrial ribosomal protein L52 |
Gene Aliases: |
- |
Location: |
Chr.14:22829860-22835037 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL52 | NM_001318870.1 | NP_001305799.1 | ||
| NM_178336.2 | NP_848026.1 | |||
| NM_180982.2 | NP_851313.1 | |||
| NM_181304.2 | NP_851821.1 | |||
| NM_181305.2 | NP_851822.1 | |||
| NM_181306.2 | NP_851823.1 | |||
| NM_181307.2 | NP_851824.1 | |||
| XM_005267325.4 | XP_005267382.1 | |||
| XM_005267328.3 | XP_005267385.1 | |||
| AA282746.1 | ||||
| AI380351.1 | ||||
| AK000450.1 | ||||
| AK096946.2 | 3 | 1660 | ||
| AK292011.1 | ||||
| BC068070.1 | AAH68070.1 | |||
| BG714882.1 | ||||
| BG716120.1 | ||||
| BQ082981.1 | ||||
| BU600884.1 | ||||
| BX248749.1 | CAD66556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain |
|
| esv3633763 | Chr.14:22686795 - 22843098 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map