Assay Details
Target Gene Details
Entrez Gene ID: | 145483 |
Gene Name: | family with sequence similarity 161 member B |
Gene Aliases: |
C14orf44, c14_5547 |
Location: |
Chr.14:73932031-73950414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM161B | NM_152445.2 | NP_689658.2 | ||
| XM_011536475.1 | XP_011534777.1 | |||
| AI082216.1 | ||||
| CR749537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| esv274934 | Chr.14:73932913 - 73939328 on Build GRCh38 | Gain+Loss |
|
| dgv663e212 | Chr.14:73931191 - 73936946 on Build GRCh38 | Loss |
|

Genomic Map