Assay Details
Target Gene Details
Entrez Gene ID: | 7158 |
Gene Name: | tumor protein p53 binding protein 1 |
Gene Aliases: |
53BP1, TDRD30, TP53, p202, p53BP1 |
Location: |
Chr.15:43403064-43510509 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP1 | XM_011521985.1 | 27 | 7012 | XP_011520287.1 |
| XM_017022533.1 | 29 | 7267 | XP_016878022.1 | |
| AK125173.1 |
Target Gene Details
Entrez Gene ID: | 27229 |
Gene Name: | tubulin gamma complex associated protein 4 |
Gene Aliases: |
76P, GCP-4, GCP4, Grip76, MCCRP3 |
Location: |
Chr.15:43371059-43409771 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBGCP4 | NM_001286414.2 | 18 | 3353 | NP_001273343.1 |
| NM_014444.4 | 18 | 3350 | NP_055259.2 | |
| XM_011521454.2 | 17 | 3236 | XP_011519756.1 | |
| XM_011521455.2 | 18 | 3251 | XP_011519757.1 | |
| XM_017022078.1 | 17 | 3233 | XP_016877567.1 |
More Information
Set Membership: |
|

Genomic Map