Assay Details
Target Gene Details
Entrez Gene ID: | 7090 |
Gene Name: | transducin like enhancer of split 3 |
Gene Aliases: |
ESG, ESG3, GRG3, HsT18976 |
Location: |
Chr.15:70047790-70097935 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLE3 | NM_001105192.2 | 20 | 4462 | NP_001098662.1 |
| NM_001282979.1 | 20 | 4447 | NP_001269908.1 | |
| NM_001282980.1 | 20 | 4456 | NP_001269909.1 | |
| NM_001282981.1 | 20 | 3395 | NP_001269910.1 | |
| NM_001282982.1 | 18 | 3202 | NP_001269911.1 | |
| NM_005078.3 | 20 | 4471 | NP_005069.2 | |
| NM_020908.2 | 20 | 4435 | NP_065959.1 | |
| XM_005254622.4 | 20 | 4510 | XP_005254679.1 | |
| XM_005254623.3 | 20 | 4495 | XP_005254680.1 | |
| XM_005254625.3 | 20 | 4483 | XP_005254682.1 | |
| XM_005254628.3 | 20 | 4468 | XP_005254685.1 | |
| XM_005254633.2 | 20 | 4438 | XP_005254690.1 | |
| XM_006720665.3 | 20 | 4462 | XP_006720728.1 | |
| XM_011521976.2 | 20 | 4519 | XP_011520278.1 | |
| XM_011521977.2 | 20 | 4516 | XP_011520279.1 | |
| XM_011521978.2 | 20 | 4504 | XP_011520280.1 | |
| XM_011521979.2 | 19 | 3310 | XP_011520281.1 | |
| XM_011521980.2 | 20 | 4486 | XP_011520282.1 | |
| XM_011521982.2 | 18 | 4267 | XP_011520284.1 | |
| XM_011521983.2 | 18 | 4252 | XP_011520285.1 | |
| AB046767.1 | 20 | 4417 | ||
| AB209672.1 | 3 | 7723 | BAD92909.1 | |
| AK000083.1 | 1 | 1037 | ||
| AK001258.1 | 9 | 2298 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951987 | Chr.15:70043962 - 70061561 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map