Assay Details
Target Gene Details
Entrez Gene ID: | 79768 |
Gene Name: | katanin regulatory subunit B1 like 1 |
Gene Aliases: |
C15orf29 |
Location: |
Chr.15:34140674-34210146 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNBL1 | NM_024713.2 | NP_078989.1 | ||
| XM_005254668.3 | XP_005254725.1 | |||
| XM_017022571.1 | XP_016878060.1 | |||
| XM_017022572.1 | XP_016878061.1 | |||
| XM_017022573.1 | XP_016878062.1 | |||
| XM_017022574.1 | XP_016878063.1 | |||
| XM_017022575.1 | XP_016878064.1 | |||
| AK026210.1 | BAB15394.1 | |||
| AK293021.1 | ||||
| AK293695.1 | ||||
| AK297904.1 | ||||
| AK311338.1 | ||||
| AL136908.1 | CAB66842.1 | |||
| AL833631.1 | ||||
| BC026234.1 | AAH26234.1 | |||
| BC111000.1 | AAI11001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain |
|
| esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss |
|
| esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain |
|
| dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147640282] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map