Assay Details
Target Gene Details
Entrez Gene ID: | 79895 |
Gene Name: | ATPase phospholipid transporting 8B4 (putative) |
Gene Aliases: |
ATPIM |
Location: |
Chr.15:49858238-50182096 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833003 | Chr.15:49808175 - 49981112 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map