Assay Details
Target Gene Details
Entrez Gene ID: | 83464 |
Gene Name: | aph-1 homolog B, gamma-secretase subunit |
Gene Aliases: |
APH-1B, PRO1328, PSFL, TAAV688 |
Location: |
Chr.15:63277519-63309126 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APH1B | NM_001145646.1 | NP_001139118.1 | ||
| NM_031301.3 | NP_112591.2 | |||
| XM_011522105.2 | XP_011520407.1 | |||
| AA808355.1 | ||||
| AB189172.1 | BAD95573.1 | |||
| AB197122.1 | BAE02660.1 | |||
| AF508794.1 | AAN63817.1 | |||
| AK291204.1 | ||||
| AK307456.1 | ||||
| AK307725.1 | ||||
| AK310843.1 | ||||
| AL136671.1 | CAB66606.1 | |||
| AY358698.1 | AAQ89061.1 | |||
| BC020905.1 | AAH20905.1 | |||
| BI825203.1 | ||||
| DA397427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428305 | Chr.15:63040203 - 63322630 on Build GRCh38 | Gain |
|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114004105] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map