Assay Details
Target Gene Details
Entrez Gene ID: | 5315 |
Gene Name: | pyruvate kinase, muscle |
Gene Aliases: |
CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1 |
Location: |
Chr.15:72199029-72231624 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833051 | Chr.15:72023130 - 72216088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map