Assay Details
Target Gene Details
Entrez Gene ID: | 5315 |
Gene Name: | pyruvate kinase, muscle |
Gene Aliases: |
CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1 |
Location: |
Chr.15:72199029-72231624 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map