Assay Details
Target Gene Details
Entrez Gene ID: | 4649 |
Gene Name: | myosin IXA |
Gene Aliases: |
- |
Location: |
Chr.15:71822290-72118577 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO9A | NM_006901.3 | 1 | 213 | NP_008832.2 |
| XM_006720539.2 | 1 | 213 | XP_006720602.1 | |
| XM_011521613.2 | 1 | 213 | XP_011519915.1 | |
| XM_011521615.2 | XP_011519917.1 | |||
| XM_011521616.2 | XP_011519918.1 | |||
| XM_011521617.2 | 1 | 213 | XP_011519919.1 | |
| XM_011521618.2 | 1 | 213 | XP_011519920.1 | |
| XM_011521619.2 | 1 | 213 | XP_011519921.1 | |
| XM_011521620.2 | 1 | 213 | XP_011519922.1 | |
| XM_011521621.2 | 1 | 213 | XP_011519923.1 | |
| XM_011521622.2 | 1 | 213 | XP_011519924.1 | |
| XM_017022229.1 | XP_016877718.1 | |||
| XM_017022230.1 | 1 | 213 | XP_016877719.1 | |
| BC060886.1 | 1 | 195 | AAH60886.1 | |
| DB085692.1 | 1 | 190 | ||
| DB100556.1 | 1 | 213 |
Target Gene Details
Entrez Gene ID: | 123228 |
Gene Name: | SUMO/sentrin peptidase family member, NEDD8 specific |
Gene Aliases: |
DEN1, NEDP1, PRSC2 |
Location: |
Chr.15:72114258-72143688 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP8 | NM_001166340.1 | NP_001159812.1 | ||
| NM_001172109.1 | NP_001165580.1 | |||
| NM_001172110.1 | NP_001165581.1 | |||
| XM_005254157.3 | XP_005254214.1 | |||
| XM_011521215.2 | XP_011519517.1 | |||
| XM_011521217.2 | XP_011519519.1 | |||
| AK097557.1 | ||||
| BP370384.1 | ||||
| DB048641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833051 | Chr.15:72023130 - 72216088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map