Assay Details
Target Gene Details
Entrez Gene ID: | 26263 |
Gene Name: | F-box protein 22 |
Gene Aliases: |
FBX22, FISTC1 |
Location: |
Chr.15:75903859-75937513 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO22 | XM_011521447.2 | 8 | 4397 | XP_011519749.1 |
Target Gene Details
Entrez Gene ID: | 145957 |
Gene Name: | neuregulin 4 |
Gene Aliases: |
HRG4 |
Location: |
Chr.15:75935190-76062997 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRG4 | XM_017021937.1 | 12 | 1324 | XP_016877426.1 |
| XM_017021938.1 | 11 | 3531 | XP_016877427.1 | |
| XM_017021939.1 | 11 | 1093 | XP_016877428.1 | |
| XM_017021940.1 | 10 | 3869 | XP_016877429.1 | |
| XM_017021941.1 | 8 | 832 | XP_016877430.1 | |
| XM_017021942.1 | 8 | 850 | XP_016877431.1 | |
| XM_017021943.1 | 8 | 923 | XP_016877432.1 | |
| XM_017021944.1 | 9 | 3745 | XP_016877433.1 | |
| XM_017021946.1 | 10 | 3820 | XP_016877435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| nsv1044733 | Chr.15:75828583 - 75936539 on Build GRCh38 | Loss |
|
| esv3636933 | Chr.15:75908231 - 76025938 on Build GRCh38 | Gain |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| dgv2622n100 | Chr.15:75904333 - 76022201 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map