Assay Details
Target Gene Details
Entrez Gene ID: | 27229 |
Gene Name: | tubulin gamma complex associated protein 4 |
Gene Aliases: |
76P, GCP-4, GCP4, Grip76, MCCRP3 |
Location: |
Chr.15:43371059-43409771 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBGCP4 | NM_001286414.2 | NP_001273343.1 | ||
| NM_014444.4 | NP_055259.2 | |||
| XM_011521454.2 | XP_011519756.1 | |||
| XM_011521455.2 | XP_011519757.1 | |||
| XM_017022078.1 | XP_016877567.1 | |||
| AJ249677.1 | CAB62539.1 | |||
| AK001639.1 | BAA91802.1 | |||
| AK026551.1 | ||||
| AK027703.1 | ||||
| BC000966.2 | AAH00966.1 | |||
| BC009870.2 | AAH09870.1 | |||
| BC012801.2 | AAH12801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1513 | Chr.15:43362170 - 43396516 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map