Assay Details
Target Gene Details
Entrez Gene ID: | 161742 |
Gene Name: | sprouty related EVH1 domain containing 1 |
Gene Aliases: |
NFLS, PPP1R147, hSpred1, spred-1 |
Location: |
Chr.15:38252087-38357249 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRED1 | NM_152594.2 | NP_689807.1 | ||
| XM_005254202.3 | XP_005254259.1 | |||
| XM_011521289.2 | XP_011519591.1 | |||
| AK091222.1 | BAC03614.1 | |||
| AY299089.1 | AAP59414.1 | |||
| BC018015.1 | ||||
| BC137480.1 | ||||
| BC137481.1 | ||||
| DA405650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892674 | Chr.15:37311235 - 38400599 on Build GRCh38 | Gain |
|
| nsv832974 | Chr.15:38206405 - 38398384 on Build GRCh38 | Loss |
|
| nsv832973 | Chr.15:38151604 - 38352681 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map