Assay Details
Target Gene Details
Entrez Gene ID: | 2562 |
Gene Name: | gamma-aminobutyric acid type A receptor beta3 subunit |
Gene Aliases: |
ECA5 |
Location: |
Chr.15:26543546-26773788 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRB3 | NM_000814.5 | NP_000805.1 | ||
| NM_001191320.1 | NP_001178249.1 | |||
| NM_001191321.2 | NP_001178250.1 | |||
| NM_001278631.1 | NP_001265560.1 | |||
| NM_021912.4 | NP_068712.1 | |||
| XM_011521428.2 | XP_011519730.1 | |||
| AK295167.1 | ||||
| AK302822.1 | ||||
| AK315311.1 | ||||
| BC010641.1 | AAH10641.1 | |||
| CD013894.1 | ||||
| KJ534842.1 | ||||
| M82919.1 | AAA52511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv144n21 | Chr.15:26513916 - 26649934 on Build GRCh38 | Gain |
|
| nsv469894 | Chr.15:26558113 - 26745817 on Build GRCh38 | Loss |
|
| nsv568598 | Chr.15:26082319 - 26575978 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map