Assay Details
Target Gene Details
Entrez Gene ID: | 54956 |
Gene Name: | poly(ADP-ribose) polymerase family member 16 |
Gene Aliases: |
ARTD15, C15orf30, pART15 |
Location: |
Chr.15:65231691-65286886 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP16 | XM_006720589.2 | XP_006720652.1 | ||
| XM_006720590.2 | XP_006720653.1 |
More Information
Set Membership: |
|

Genomic Map