Assay Details
Target Gene Details
Entrez Gene ID: | 80125 |
Gene Name: | coiled-coil domain containing 33 |
Gene Aliases: |
CT61, HP11097 |
Location: |
Chr.15:74202705-74336472 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC33 | XM_011522085.2 | 1 | 68 | XP_011520387.1 |
| XM_017022623.1 | 1 | 69 | XP_016878112.1 | |
| XM_017022624.1 | 1 | 68 | XP_016878113.1 | |
| XM_017022625.1 | 1 | 68 | XP_016878114.1 | |
| XM_017022626.1 | 1 | 68 | XP_016878115.1 | |
| XM_017022627.1 | 1 | 68 | XP_016878116.1 | |
| XM_017022628.1 | 1 | 68 | XP_016878117.1 | |
| XM_017022629.1 | XP_016878118.1 | |||
| XM_017022630.1 | 1 | 70 | XP_016878119.1 | |
| XM_017022631.1 | 1 | 69 | XP_016878120.1 | |
| XM_017022633.1 | 1 | 78 | XP_016878122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3351294 | Chr.15:74216501 - 74216728 on Build GRCh38 | Duplication |
|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv471253 | Chr.15:74196866 - 74252807 on Build GRCh38 | Loss |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| esv3451538 | Chr.15:74216410 - 74216850 on Build GRCh38 | Duplication |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map