Assay Details
Target Gene Details
Entrez Gene ID: | 10363 |
Gene Name: | high mobility group 20A |
Gene Aliases: |
HMGX1, HMGXB1 |
Location: |
Chr.15:77420651-77519899 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMG20A | NM_001304504.1 | NP_001291433.1 | ||
| NM_001304505.1 | NP_001291434.1 | |||
| NM_018200.3 | NP_060670.1 | |||
| XM_011521158.2 | XP_011519460.1 | |||
| AF146222.1 | AAF66706.1 | |||
| AK001601.1 | BAA91782.1 | |||
| AK096546.1 | ||||
| AK223100.1 | BAD96820.1 | |||
| AK297449.1 | ||||
| AK311376.1 | ||||
| BC021959.1 | AAH21959.1 | |||
| DA333471.1 | ||||
| DB068283.1 | ||||
| DB213761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map