Assay Details
Target Gene Details
Entrez Gene ID: | 123591 |
Gene Name: | transmembrane protein 266 |
Gene Aliases: |
C15orf27, HVRP1 |
Location: |
Chr.15:76059929-76204963 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM266 | NM_152335.2 | NP_689548.2 | ||
| XM_005254160.2 | XP_005254217.1 | |||
| XM_017021915.1 | XP_016877404.1 | |||
| XM_017021916.1 | XP_016877405.1 | |||
| XM_017021917.1 | XP_016877406.1 | |||
| AK056777.1 | ||||
| AK095509.1 | BAC04562.1 | |||
| AK308676.1 | ||||
| DA090797.1 | ||||
| DA483185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| nsv1048760 | Chr.15:76042203 - 76204636 on Build GRCh38 | Gain |
|
| nsv569998 | Chr.15:75940264 - 76258597 on Build GRCh38 | Gain |
|
| esv2669418 | Chr.15:76073911 - 76078466 on Build GRCh38 | Deletion |
|
| nsv833057 | Chr.15:76075507 - 76293527 on Build GRCh38 | Loss |
|
| esv3636935 | Chr.15:76073935 - 76078476 on Build GRCh38 | Loss |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76631432] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map