Assay Details
Target Gene Details
Entrez Gene ID: | 23604 |
Gene Name: | death associated protein kinase 2 |
Gene Aliases: |
DRP-1, DRP1 |
Location: |
Chr.15:63907036-64046470 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Gene Details
Entrez Gene ID: | 101928988 |
Gene Name: | uncharacterized LOC101928988 |
Gene Aliases: |
- |
Location: |
Chr.15:63928274-63935953 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928988 | NR_120344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636720 | Chr.15:63860455 - 64010781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map